accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NN

CAS RN 826-85-7

IUPAC Name: 1-phenyl-1H-pyrazol-5-amine
Synonyms: pyrazole, 5-amino-1-phenyl-
Molecular Weight (g/mol): 159.19
Molecular Formula: C9H9N3
InChi Key: ZVNYYNAAEVZNDW-UHFFFAOYSA-N
SMILES: NC1=CC=NN1C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results