accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNH2NCl

CAS RN 82632-77-7

IUPAC Name: 4-(4-chlorophenyl)-5-methyl-1,3-thiazol-2-amine
Synonyms: 4-(4-chlorophenyl)-5-methyl-1,3-thiazol-2-amine
Molecular Weight (g/mol): 224.71
Molecular Formula: C10H9ClN2S
InChi Key: IEJGVSUOFMBPJU-UHFFFAOYSA-N
SMILES: CC1=C(N=C(N)S1)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results