accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOClFFFH3CCH3(Z)

CAS RN 82657-04-3

IUPAC Name: {2-methyl-[1,1'-biphenyl]-3-yl}methyl 3-[(1Z)-2-chloro-3,3,3-trifluoroprop-1-en-1-yl]-2,2-dimethylcyclopropane-1-carboxylate
Synonyms: talstar epa pesticide chemical code 128825 biphentrin caswell no. 463F bifenthrine biphenate brigade capture
Molecular Weight (g/mol): 422.87
Molecular Formula: C23H22ClF3O2
InChi Key: OMFRMAHOUUJSGP-UNOMPAQXNA-N
SMILES: CC1=C(COC(=O)C2C(\C=C(/Cl)C(F)(F)F)C2(C)C)C=CC=C1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results