accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 82657-72-5

IUPAC Name: 1-(bromomethyl)-2-phenoxybenzene
Synonyms: 1-(bromomethyl)-2-phenoxybenzene
Molecular Weight (g/mol): 263.13
Molecular Formula: C13H11BrO
InChi Key: YQRIQBOWLXRKKG-UHFFFAOYSA-N
SMILES: BrCC1=CC=CC=C1OC1=CC=CC=C1
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results