accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFBrBrF

CAS RN 827-08-7

IUPAC Name: 1,2-dibromo-3,4,5,6-tetrafluorobenzene
Synonyms: 1,2-dibromo-3,4,5,6-tetrafluorobenzene
Molecular Weight (g/mol): 307.87
Molecular Formula: C6Br2F4
InChi Key: IPLUWQPTPKNBRD-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(Br)C(Br)=C1F
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)