accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2BrBrNOO

CAS RN 827-94-1

IUPAC Name: 2,6-dibromo-4-nitroaniline
Synonyms: 2,6-dibromo-4-nitroaniline
Molecular Weight (g/mol): 295.92
Molecular Formula: C6H4Br2N2O2
InChi Key: YMZIFDLWYUSZCC-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(C=C1Br)[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results