CAS RN 82769-76-4
IUPAC Name:
3-amino-3-phenylpropan-1-ol
Synonyms:
3-amino-3-phenylpropan-1-ol
Molecular Weight (g/mol):
151.21
Molecular Formula:
C9H13NO
InChi Key:
SEQXIQNPMQTBGN-UHFFFAOYNA-N
SMILES:
NC(CCO)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
(S)-3-Amino-3-phenylpropan-1-ol, 95%, 98% ee, Thermo Scientific Chemicals
(S)-3-Amino-3-phenylpropan-1-ol, 95%, 98% ee, Thermo Scientific Chemicals
CAS: 82769-76-4 Molecular Formula: C9H13NO Molecular Weight (g/mol): 151.21 MDL Number: MFCD01311768 InChI Key: SEQXIQNPMQTBGN-VIFPVBQESA-N Synonym: s-3-amino-3-phenylpropan-1-ol,3s-3-amino-3-phenylpropan-1-ol,s-1-phenyl-3-propanolamine,s-beta-phenylalaninol,s-3-phenyl-beta-alaninol,s-3-amino-3-phenyl-1-propanol,s-3-amino-3-phenyl-propan-1-ol,3s-3-amino-3-phenyl-propan-1-ol,pubchem13854,ksc491q5n PubChem CID: 2734520 IUPAC Name: (3S)-3-amino-3-phenylpropan-1-ol SMILES: C1=CC=C(C=C1)C(CCO)N