accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 82777-09-1

IUPAC Name: 2-iodo-3-phenyl-1,1'-biphenyl
Synonyms: 2-iodo-3-phenyl-1,1'-biphenyl
Molecular Weight (g/mol): 356.21
Molecular Formula: C18H13I
InChi Key: RLZYBGOJAWOQMK-UHFFFAOYSA-N
SMILES: IC1=C(C=CC=C1C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results