accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNHFFFFF

CAS RN 828-73-9

IUPAC Name: (2,3,4,5,6-pentafluorophenyl)hydrazine
Synonyms: (2,3,4,5,6-pentafluorophenyl)hydrazine
Molecular Weight (g/mol): 198.1
Molecular Formula: C6H3F5N2
InChi Key: BYCUWCJUPSUFBX-UHFFFAOYSA-N
SMILES: NNC1=C(F)C(F)=C(F)C(F)=C1F
Molecular Weight (g/mol) 
  • (1)
  • (6)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (6)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)