CAS RN 829-85-6
CAS RN 829-85-6
IUPAC Name:
diphenylphosphane
Synonyms:
diphenylphosphine
Molecular Weight (g/mol):
186.19
Molecular Formula:
C12H11P
InChi Key:
GPAYUJZHTULNBE-UHFFFAOYSA-N
SMILES:
P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
2
of
2
results
1
Sigma Aldrich Diphenylphosphine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
2
Thermo Scientific Chemicals Diphenylphosphine
CAS: 829-85-6 Molecular Formula: C12H11P Molecular Weight (g/mol): 186.19 MDL Number: MFCD00003040 InChI Key: GPAYUJZHTULNBE-UHFFFAOYSA-N Synonym: diphenylphosphine,phosphine, diphenyl,ph2ph,unii-f9b5t7o7zy,diphenyl phosphine,hpph2,f9b5t7o7zy,diphenylphosphine 10wt%inhexane,diphenylphospine,diphenyl-phosphine PubChem CID: 70017 IUPAC Name: diphenylphosphane SMILES: P(C1=CC=CC=C1)C1=CC=CC=C1