accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 82911-71-5

IUPAC Name: 1H-1,2,3-benzotriazol-1-yl (9H-fluoren-9-yl)methyl carbonate
Synonyms: 1,2,3-benzotriazol-1-yl 9H-fluoren-9-ylmethyl carbonate
Molecular Weight (g/mol): 357.37
Molecular Formula: C21H15N3O3
InChi Key: KNSPZTVPSAKHBN-UHFFFAOYSA-N
SMILES: O=C(OCC1C2=CC=CC=C2C2=CC=CC=C12)ON1N=NC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results