CAS RN 82911-79-3
IUPAC Name:
methyl (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-hydroxyphenyl)propanoate
Synonyms:
methyl (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
Molecular Weight (g/mol):
417.46
Molecular Formula:
C25H23NO5
InChi Key:
NWIXWDJMIUGINH-QHCPKHFHSA-N
SMILES:
COC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
1
–
1
of
1
results
1
Fmoc-Tyr-OMe, Advanced ChemTech
CAS: 82911-79-3 Molecular Formula: C25H23NO5 Molecular Weight (g/mol): 417.46 MDL Number: MFCD16037899 InChI Key: NWIXWDJMIUGINH-QHCPKHFHSA-N IUPAC Name: methyl (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-hydroxyphenyl)propanoate SMILES: COC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2