accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3H3CNOONOCH3H3CH3CCH3O(?)(?)

CAS RN 82953-57-9

IUPAC Name: 7,18-bis[2,6-bis(propan-2-yl)phenyl]-7,18-diazaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Synonyms: 7,18-bis(2,6-diisopropylphenyl)-7,18-diazaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Molecular Weight (g/mol): 710.87
Molecular Formula: C48H42N2O4
InChi Key: NAZODJSYHDYJGP-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC(C(C)C)=C1N1C(=O)C2=CC=C3C4=CC=C5C(=O)N(C(=O)C6=CC=C(C7=CC=C(C1=O)C2=C37)C4=C56)C1=C(C=CC=C1C(C)C)C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results