accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 82954-89-0

IUPAC Name: 2-ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 156.03
Molecular Formula: C8H17BO2
InChi Key: NPUBDPDASOEIOA-UHFFFAOYSA-N
SMILES: CCB1OC(C)(C)C(C)(C)O1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (3)