accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSiNHSiOH3COCH3OH3COH3COH3C

CAS RN 82985-35-1

IUPAC Name: 3,3,11,11-tetramethoxy-2,12-dioxa-7-aza-3,11-disilatridecane
Synonyms: bis(trimethoxysilylpropyl)amine
Molecular Weight (g/mol): 341.55
Molecular Formula: C12H31NO6Si2
InChi Key: TZZGHGKTHXIOMN-UHFFFAOYSA-N
SMILES: CO[Si](CCCNCCC[Si](OC)(OC)OC)(OC)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results