accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClON

CAS RN 83-01-2

IUPAC Name: N,N-diphenylcarbamoyl chloride
Synonyms: diphenylcarbamyl chloride
Molecular Weight (g/mol): 231.68
Molecular Formula: C13H10ClNO
InChi Key: XNBKKRFABABBPM-UHFFFAOYSA-N
SMILES: ClC(=O)N(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (4)
  • (4)
  • (2)
  • (6)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)