accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 83-07-8

IUPAC Name: 4-amino-1,5-dimethyl-2-phenyl-2,3-dihydro-1H-pyrazol-3-one
Synonyms: aminoantipyrine solnapyrin-A metapirazone aminoazophenazone 4-aminoantipyrene ampyrone aminoantipyrin 4-aap solvapyrin-A
Molecular Weight (g/mol): 203.25
Molecular Formula: C11H13N3O
InChi Key: RLFWWDJHLFCNIJ-UHFFFAOYSA-N
SMILES: CN1N(C(=O)C(N)=C1C)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (12)
  • (4)
Quantity 
  • (1)
  • (6)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (4)
Physical Form 
  • (5)
  • (4)
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (1)
  • (2)
  • (2)
  • (5)
  • (7)
  • (2)
  • (12)
  • (4)
  • (1)
  • (6)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (4)
  • (5)
  • (4)
  • (2)
  • (1)
  • (1)