accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 83-42-1

IUPAC Name: 1-chloro-2-methyl-3-nitrobenzene
Synonyms: 1-chloro-2-methyl-3-nitrobenzene
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: XCSNRORTQRKCHB-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=CC=C1[N+]([O-])=O
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)