accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 83-44-3

IUPAC Name: (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Synonyms: deoxycholic acid|desoxycholic acid|desoxycholsaeure|7α-deoxycholic acid
Molecular Weight (g/mol): 392.58
Molecular Formula: C24H40O4
InChi Key: KXGVEGMKQFWNSR-LLQZFEROSA-N
SMILES: [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)