accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 83-66-9

IUPAC Name: 1-tert-butyl-2-methoxy-4-methyl-3,5-dinitrobenzene
Synonyms: 1-tert-butyl-2-methoxy-4-methyl-3,5-dinitrobenzene
Molecular Weight (g/mol): 268.27
Molecular Formula: C12H16N2O5
InChi Key: SUAUILGSCPYJCS-UHFFFAOYSA-N
SMILES: COC1=C(C=C(C(C)=C1[N+]([O-])=O)[N+]([O-])=O)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results