accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3OCH3OCH3(E/Z)

CAS RN 83088-26-0

IUPAC Name: 4-[2-(3,5-dimethoxyphenyl)ethenyl]-1,2-dimethoxybenzene
Synonyms: 4-[2-(3,5-dimethoxyphenyl)ethenyl]-1,2-dimethoxybenzene
Molecular Weight (g/mol): 300.35
Molecular Formula: C18H20O4
InChi Key: PTVAOGIYBMTHSN-UHFFFAOYSA-N
SMILES: COC1=CC(C=CC2=CC(OC)=C(OC)C=C2)=CC(OC)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results