accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 831-61-8

IUPAC Name: ethyl 3,4,5-trihydroxybenzoate
Synonyms: ethyl gallate
Molecular Weight (g/mol): 198.17
Molecular Formula: C9H10O5
InChi Key: VFPFQHQNJCMNBZ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(O)=C(O)C(O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (7)
Percent Purity 
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (4)
  • (3)
  • (1)
  • (7)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)