CAS RN 831-81-2
IUPAC Name:
1-benzyl-4-chlorobenzene
Synonyms:
(4-benzyl)chlorobenzene
Molecular Weight (g/mol):
202.68
Molecular Formula:
C13H11Cl
InChi Key:
NPOGRKGIBGKRNI-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(CC2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
4-Chlorodiphenylmethane 96.0+%, TCI America™
CAS: 831-81-2 Molecular Formula: C13H11Cl Molecular Weight (g/mol): 202.681 MDL Number: MFCD00094070 InChI Key: NPOGRKGIBGKRNI-UHFFFAOYSA-N PubChem CID: 13253 IUPAC Name: 1-benzyl-4-chlorobenzene SMILES: C1=CC=C(C=C1)CC2=CC=C(C=C2)Cl