accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOHOOCH3H3CS(R)(R)NHO(R)NH2ON(S)OOOO(S)N(R)OSOOCH3CH3

CAS RN 83105-70-8

IUPAC Name: 4-methylbenzene-1-sulfonic acid; [(2S,5R,6R)-6-[(2R)-2-amino-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carbonyloxy]methyl (2S,5R)-3,3-dimethyl-4,4,7-trioxo-4λ⁶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Synonyms: para-toluene sulfonate; sultamicillin
Molecular Weight (g/mol): 766.85
Molecular Formula: C32H38N4O12S3
InChi Key: FFCSPKNZHGIDQM-CGAOXQFVSA-N
SMILES: CC1=CC=C(C=C1)S(O)(=O)=O.CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)C3=CC=CC=C3)C(=O)N2[C@H]1C(=O)OCOC(=O)[C@@H]1N2[C@@H](CC2=O)S(=O)(=O)C1(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results