accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClFClFNH2

CAS RN 83121-15-7

IUPAC Name: 3,5-dichloro-2,4-difluoroaniline
Synonyms: 3,5-dichloro-2,4-difluoroaniline
Molecular Weight (g/mol): 197.99
Molecular Formula: C6H3Cl2F2N
InChi Key: KLECNQGLBJHVSH-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=C(F)C(Cl)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results