accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 832-69-9

IUPAC Name: 1-methylphenanthrene
Synonyms: 1-methylphenanthrene
Molecular Weight (g/mol): 192.26
Molecular Formula: C15H12
InChi Key: DOWJXOHBNXRUOD-UHFFFAOYSA-N
SMILES: CC1=C2C=CC3=CC=CC=C3C2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results