accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHNOHO

CAS RN 832-97-3

IUPAC Name: 2-hydroxy-3-(1H-indol-3-yl)propanoic acid
Synonyms: indole-3-lactic acid 5-iHIPA indolelactate indole-3-lactate indolelactic acid
Molecular Weight (g/mol): 205.21
Molecular Formula: C11H11NO3
InChi Key: XGILAAMKEQUXLS-UHFFFAOYNA-N
SMILES: OC(CC1=CNC2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results