accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONH3CBOH3CH3CH3CCH3O

CAS RN 832114-00-8

IUPAC Name: 3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole
Synonyms: 3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole
Molecular Weight (g/mol): 223.08
Molecular Formula: C11H18BNO3
InChi Key: CVLHETBAROWASE-UHFFFAOYSA-N
SMILES: CC1=C(B2OC(C)(C)C(C)(C)O2)C(C)=NO1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)