accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNNHCH3H3CNSH3CN

CAS RN 834-12-8

IUPAC Name: N2-ethyl-6-(methylsulfanyl)-N4-(propan-2-yl)-1,3,5-triazine-2,4-diamine
Synonyms: ametrine geshpax epa pesticide chemical code 080801 caswell no. 431 topazol doruplant amephyt Evik ametryne, 14c-labeled gesapax
Molecular Weight (g/mol): 227.33
Molecular Formula: C9H17N5S
InChi Key: RQVYBGPQFYCBGX-UHFFFAOYSA-N
SMILES: CCNC1=NC(NC(C)C)=NC(SC)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results