CAS RN 834-24-2
IUPAC Name:
4-[(1E)-2-phenylethenyl]aniline
Synonyms:
4-[(1E)-2-phenylethenyl]aniline
Molecular Weight (g/mol):
195.27
Molecular Formula:
C14H13N
InChi Key:
VFPLSXYJYAKZCT-VOTSOKGWSA-N
SMILES:
NC1=CC=C(\C=C\C2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
4-Aminostilbene 97.0+%, TCI America™
CAS: 834-24-2 Molecular Formula: C14H13N Molecular Weight (g/mol): 195.27 MDL Number: MFCD00025377 InChI Key: VFPLSXYJYAKZCT-VOTSOKGWSA-N Synonym: 4-Stilbenamine PubChem CID: 1272788 IUPAC Name: 4-[(1E)-2-phenylethenyl]aniline SMILES: NC1=CC=C(\C=C\C2=CC=CC=C2)C=C1