accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 83435-58-9

IUPAC Name: tert-butyl (2R)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
Synonyms: tert-butyl (2R)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
Molecular Weight (g/mol): 201.27
Molecular Formula: C10H19NO3
InChi Key: BFFLLBPMZCIGRM-MRVPVSSYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC[C@@H]1CO
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)