accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 83493-63-4

IUPAC Name: 2-(4-ethoxyphenyl)-2-methylpropan-1-ol
Synonyms: 2-(4-ethoxyphenyl)-2-methylpropan-1-ol
Molecular Weight (g/mol): 194.27
Molecular Formula: C12H18O2
InChi Key: OZEZBKUHAGFQME-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1)C(C)(C)CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results