accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFNH2OOH

CAS RN 83506-93-8

IUPAC Name: 2-amino-4,5-difluorobenzoic acid
Synonyms: 2-amino-4,5-difluorobenzoic acid
Molecular Weight (g/mol): 173.12
Molecular Formula: C7H5F2NO2
InChi Key: DGOZIZVTANAGCA-UHFFFAOYSA-N
SMILES: NC1=CC(F)=C(F)C=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results