accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOHHO

CAS RN 83511-07-3

IUPAC Name: 3,7-dihydroxynaphthalene-2-carboxylic acid
Synonyms: 3,7-dihydroxynaphthalene-2-carboxylic acid
Molecular Weight (g/mol): 204.18
Molecular Formula: C11H8O4
InChi Key: QMWOUSYSNFCKAZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=CC(O)=CC=C2C=C1O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results