accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONH

CAS RN 836-30-6

IUPAC Name: 4-nitro-N-phenylaniline
Synonyms: 4-nitro-N-phenylbenzenamine
Molecular Weight (g/mol): 214.22
Molecular Formula: C12H10N2O2
InChi Key: XXYMSQQCBUKFHE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(NC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)