accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 836-42-0

IUPAC Name: 1-(benzyloxy)-4-(chloromethyl)benzene
Synonyms: 1-(benzyloxy)-4-(chloromethyl)benzene
Molecular Weight (g/mol): 232.71
Molecular Formula: C14H13ClO
InChi Key: UYQPSKUPEXAQRJ-UHFFFAOYSA-N
SMILES: ClCC1=CC=C(OCC2=CC=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)