accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNNNH

CAS RN 837-52-5

IUPAC Name: 7-chloro-4-(piperazin-1-yl)quinoline
Synonyms: 7-chloro-4-(piperazin-1-yl)quinoline
Molecular Weight (g/mol): 247.73
Molecular Formula: C13H14ClN3
InChi Key: DNXNPMDUDGUXOB-UHFFFAOYSA-N
SMILES: ClC1=CC=C2C(=C1)N=CC=C2N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results