accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 83701-22-8

IUPAC Name: 2,6-diamino-4-(piperidin-1-yl)-1-(sulfonatooxy)pyrimidin-1-ium
Synonyms: 2,6-diamino-4-(piperidin-1-yl)-1-(sulfonatooxy)pyrimidin-1-ium
Molecular Weight (g/mol): 289.31
Molecular Formula: C9H15N5O4S
InChi Key: OEOLOEUAGSPDLT-UHFFFAOYSA-N
SMILES: NC1=CC(=NC(N)=[N+]1OS([O-])(=O)=O)N1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)