accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 83724-41-8

IUPAC Name: tris(2-methoxyphenyl)bismuthane
Synonyms: tris(2-methoxyphenyl)bismuthane
Molecular Weight (g/mol): 530.38
Molecular Formula: C21H21BiO3
InChi Key: VFWRGMGLLNCHIA-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1[Bi](C1=CC=CC=C1OC)C1=CC=CC=C1OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results