accessibility menu, dialog, popup

UserName

Viewing profile as user:
NFIF

CAS RN 837364-88-2

IUPAC Name: 2,4-difluoro-3-iodopyridine
Synonyms: 2,4-difluoro-3-iodopyridine
Molecular Weight (g/mol): 240.98
Molecular Formula: C5H2F2IN
InChi Key: AYKASHJPLZSRDC-UHFFFAOYSA-N
SMILES: FC1=C(I)C(F)=NC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results