accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 837376-47-3

IUPAC Name: 1-[5-(pyridin-3-yl)thiophen-2-yl]methanamine
Synonyms: 1-[5-(pyridin-3-yl)thiophen-2-yl]methanamine
Molecular Weight (g/mol): 190.26
Molecular Formula: C10H10N2S
InChi Key: JHLIGYPHPBLDDL-UHFFFAOYSA-N
SMILES: NCC1=CC=C(S1)C1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results