accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOCH3CH3CH3CH3HNO

CAS RN 837392-64-0

IUPAC Name: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-indol-2-one
Synonyms: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-dihydroindol-2-one
Molecular Weight (g/mol): 259.11
Molecular Formula: C14H18BNO3
InChi Key: BXFPTCYBFJOZHJ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=C(NC(=O)C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)