accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 837392-67-3

IUPAC Name: tert-butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-indole-1-carboxylate
Synonyms: tert-butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydroindole-1-carboxylate
Molecular Weight (g/mol): 345.25
Molecular Formula: C19H28BNO4
InChi Key: OOORQXGLIKPNDK-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC2=C1C=CC(=C2)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results