accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 83799-54-6

IUPAC Name: 4-(chloromethyl)-4'-methyl-2,2'-bipyridine
Synonyms: 4-(chloromethyl)-4'-methyl-2,2'-bipyridine
Molecular Weight (g/mol): 218.68
Molecular Formula: C12H11ClN2
InChi Key: UHTKTJKJDJOOKE-UHFFFAOYSA-N
SMILES: CC1=CC(=NC=C1)C1=NC=CC(CCl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results