accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 838-85-7

IUPAC Name: diphenoxyphosphinic acid
Synonyms: diphenyl phosphate
Molecular Weight (g/mol): 250.19
Molecular Formula: C12H11O4P
InChi Key: ASMQGLCHMVWBQR-UHFFFAOYSA-N
SMILES: OP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (4)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (4)
  • (3)
  • (4)