accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 83846-48-4

CH3ICl

CAS RN 83846-48-4

IUPAC Name: 2-chloro-4-iodo-1-methylbenzene
Synonyms: 2-chloro-4-iodo-1-methylbenzene
Molecular Weight (g/mol): 252.48
Molecular Formula: C7H6ClI
InChi Key: PJYASWQMTNNSSL-UHFFFAOYSA-N
SMILES: CC1=CC=C(I)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results