accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SO

CAS RN 83846-85-9

IUPAC Name: {4-[(4-methylphenyl)sulfanyl]phenyl}(phenyl)methanone
Synonyms: {4-[(4-methylphenyl)sulfanyl]phenyl}(phenyl)methanone
Molecular Weight (g/mol): 304.41
Molecular Formula: C20H16OS
InChi Key: DBHQYYNDKZDVTN-UHFFFAOYSA-N
SMILES: CC1=CC=C(SC2=CC=C(C=C2)C(=O)C2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results