accessibility menu, dialog, popup

UserName

Viewing profile as user:
HHClClHOOONNCl

CAS RN 83881-52-1

IUPAC Name: dihydrogen 2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetic acid dichloride
Synonyms: dihydrogen cetirizine dichloride
Molecular Weight (g/mol): 461.81
Molecular Formula: C21H27Cl3N2O3
InChi Key: PGLIUCLTXOYQMV-UHFFFAOYNA-N
SMILES: [H+].[H+].[Cl-].[Cl-].OC(=O)COCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)