accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOFF

CAS RN 83882-67-1

IUPAC Name: 1-[4-(difluoromethoxy)phenyl]ethan-1-one
Synonyms: 1-[4-(difluoromethoxy)phenyl]ethanone
Molecular Weight (g/mol): 186.16
Molecular Formula: C9H8F2O2
InChi Key: GIGWRVLNOYPOIT-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(OC(F)F)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results