accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 83883-25-4

IUPAC Name: 4-[(6-hydroxyhexyl)oxy]benzoic acid
Synonyms: 4-[(6-hydroxyhexyl)oxy]benzoic acid
Molecular Weight (g/mol): 238.28
Molecular Formula: C13H18O4
InChi Key: VVYHWYFUTOHXRH-UHFFFAOYSA-N
SMILES: OCCCCCCOC1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results